Anti-CD28 Mouse Monoclonal Antibody (CF594) [clone: C28/77]
Nome del fornitore: Biotium
Certificati
Informazioni su questo articolo
Recognizes a glycoprotein of 44-88 kDa, which is identified as CD28. It is the critical T-cell co-stimulatory receptor which provides to the cell the important second activation signal by binding CD80 and CD86 that are expressed by antigen presenting cells. Besides its co-stimulation role, CD28 functions in preventing T-cells from anergic hyporesponsive state or from undergoing premature apoptotic cell death. CD28 is also expressed on human fetal NK cells and some NK cell lines, whereas on murine NK cells the CD28 expression is much broader.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: C28/77
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifiche
- Catalog No:
- BNC940077-500
- BNC940077-100
- Antigen symbol:
- CD28
- CD28
- Antigen synonyms:
- Tp44|T-cell-specific surface glycoprotein CD28
- Tp44|T-cell-specific surface glycoprotein CD28
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 594
- CF® 594
- Clone:
- Clone C28/77
- Clone C28/77
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 591629
- 591629
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifiche
Frequently Bought Together