Order Entry
Italy
ContactUsLinkComponent
Anti-CD28 Mouse Monoclonal Antibody (Biotin) [clone: C28/77]
Anti-CD28 Mouse Monoclonal Antibody (Biotin) [clone: C28/77]
Supplier:  Biotium
Certificates
Anti-CD28 Mouse Monoclonal Antibody (Biotin) [clone: C28/77]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB0077-500
  • BNCB0077-100
  • Antigen symbol:
  • CD28
  • CD28
  • Antigen synonyms:
  • Tp44|T-cell-specific surface glycoprotein CD28
  • Tp44|T-cell-specific surface glycoprotein CD28
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone C28/77
  • Clone C28/77
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 591629
  • 591629
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together