Order Entry
Italy
ContactUsLinkComponent
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (Biotin) [clone: C-51]
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (Biotin) [clone: C-51]
Nome del fornitore:  Biotium
Certificati
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (Biotin) [clone: C-51]
Nome del fornitore:  Biotium

Specifiche

  • Catalog No:
  • BNCB0034-500
  • BNCB0034-100
  • Antigen symbol:
  • KRT8 & KRT18
  • KRT8 & KRT18
  • Antigen name:
  • Cytokeratin 8/18
  • Cytokeratin 8/18
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone C-51
  • Clone C-51
  • Reactivity:
  • Pig, Monkey, Sheep, Human, Bovine
  • Pig, Monkey, Sheep, Human, Bovine
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 533782 , 708445 (CK8); 406013 (CK18)
  • 533782 , 708445 (CK8); 406013 (CK18)
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifiche

Frequently Bought Together