Order Entry
Italy
ContactUsLinkComponent
Anti-FN1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: TV-1]
Anti-FN1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: TV-1]
Catalog # BNUM0029-50
Supplier:  Biotium
Anti-FN1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: TV-1]
Catalog # BNUM0029-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibronectin
  • Antigen symbol:
    FN1
  • Clonality:
    Monoclonal
  • Clone:
    Clone TV-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Pig,
    Human,
    Rat,
    Mouse
  • Gene ID:
    203717
  • Antigen synonyms:
    FN1|Ugl-Y3|FNZ|Migration stimulating factor (MSF)|GFND2|Cold insoluble globulin (CIG)|LETS|GFND|FINC
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

Fibronectin is a soluble dimeric glycoprotein of 440 kDa, which is present in cells, extracellular matrix, and blood. This MAb reacts with the cellular as well as plasma form of fibronectin. Reportedly, after iv administration, this MAb localizes to tumor vessels where it binds to the underlying basement. Epitope recognized by this antibody is not accessible in normal tissues to the circulating MAb indicating that it can be used to specifically target tumor vessels in vivo. TV-1 is reportedly useful for delivering vasoactive agents to tumors to induce increased vascular permeability or blood flow prior to treatment with chemotherapeutic drugs or MAbs.

Type: Primary
Antigen: FN1
Clonality: Monoclonal
Clone: TV-1
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Pig, Rat