Analytical HPLC columns, Ultra silica (USP L3)
Supplier: Restek
Certificates
About this item
Ultra silica columns made of high-purity, type-B silica are designed for selective and reliable HPLC applications.
- Base-deactivated spherical silica
- Minimises activity and creates high-density bonding
Frequently Bought Together