Inserisci ordine
Italy
ContactUsLinkComponent
 
Anti-UXT Goat Polyclonal Antibody (Biotinylated)
Catalog # EVBIEB08231-B
Supplier:  Everest Biotech
Anti-UXT Goat Polyclonal Antibody (Biotinylated)
Catalog # EVBIEB08231-B
Supplier:  Everest Biotech

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Ubiquitously-expressed Transcript
  • Simbolo dell'antigene:
    UXT
  • Clonalità:
    Policlonale
  • Coniugazione:
    Biotina
  • ELISA:
    Yes
  • Ospite:
    Goat
  • ImmunoChimica:
    Yes
  • Isotipo:
    IgG
  • Reattività:
    Human
  • Western blot:
    Yes
  • Formato:
    Affinity Purified
  • Forma:
    Liquid
  • ID gene:
    8409
  • Sinomini antigene:
    ubiquitously expressed transcript protein|SKP2-associated alpha PFD 1|ART-27|ubiquitously expressed prefoldin like chaperone|prefoldin-like chaperone|UXT|ubiquitously-expressed|STAP1|androgen receptor trapped clone 27 protein
  • Tampone di conservazione:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
  • Sequenza:
    C-DHRDKVYEQLAKY.
  • Temperatura di conservazione:
    +8 °C
  • Concentrazione:
    0,5 µg/µl
  • Immunogeno:
    UXT, Biotinylated
  • Taglia:
    200 μL
  • Tipo di conf.:
    Plastic Vial
  • Conf:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: UXT
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human