Order Entry
Italy
ContactUsLinkComponent
1,3-Diolein ~99%, frozen liquid
1,3-Diolein ~99%, frozen liquid
Catalog # ICNA0219515210
Supplier:  MP Biomedicals
CAS Number:  
1,3-Diolein ~99%, frozen liquid
Catalog # ICNA0219515210
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ~99%

Specification Test Results

About this item

 :
 :621 g/mol
Storage Temperature:Freezer
CAS Number:2465-32-9
Formula:C₃₉H₇₂O₅
MDL Number:MFCD00042913