Ammonium acetate, anhydrous, powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:631-61-8
:
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
:77.08 g/mol
Specification Test Results
- Synonyms:Acetic acid ammonium salt
- Form:White Crystalline Powder
- Storage:Store at Room Temperature (15 - 30 °C), Desiccate
- Assay:≥97%
- Heavy Metals:≤5 ppm
- Insoluble Matter:≤0,005%
- pH:6,7 - 7,3 (5% aq soln)
- Residue on Ignition:≤0,01%
- Chloride (Cl):≤5 ppm
- Nitrate (NO3):0,001%
- Sulfate (SO4):≤0,001%
- Iron (Fe):≤5 ppm
Specification Test Results
Frequently Bought Together