Solvent blue 35, dark red/purple almost black powder C.I. 61554
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:17354-14-2
:
Formula:C₂₂H₂₆N₂O₂
MDL Number:MFCD00011714
:350.45 g/mol
Melting Pt:120…122 °C (lit.)
Density:1.23 g/mL (lit.)
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
Specification Test Results
Frequently Bought Together