Primaquine diphosphate, dark orange powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:63-45-6
:
Formula:C₁₅H₂₇N₃O₉P₂
MDL Number:MFCD00013166
:455.33 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together