Neohesperidin, off-white powder
Catalog # ICNA0215581380
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:610.60 g/mol
Storage Temperature:Refrigerator
CAS Number:13241-33-3
Formula:C₂₈H₃₄O₁₅
MDL Number:MFCD16661259