Fmoc-Tle-OH ≥99.0% (by HPLC), Novabiochem®
: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tert-leucine, (S)-2-(Fmoc-amino)-3,3-dimethylbutyric acid, Fmoc-Tle-OH, Fmoc-α-t-butylglycine, (S)-2-[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]-3,3-dimethylbutanoic acid, N-Fmoc-L-tert-leucine, Fmoc-tBu-Gly-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3,3-dimethylbutanoic acid
:132684-60-7
:
:C₂₁H₂₃NO₄
:MFCD00065649
:124…127 °C
:353.418 g/mol
- Environmentally Preferable:
- Alternate Names:N-α-Fmoc-L-α-t.-butyl-glycine, Fmoc-Tle-OH
- Colour (visual):White to slight yellow to beige
- Appearance of substance (visual):Powder
- Identity (IR):Passes Test
- Enantiomeric purity:≥99,5% (a/a)
- Purity (TLC(011A)):≥98%
- Purity (TLC(0811)):≥98%
- Assay (HPLC, area%):≥99,0% (a/a)
- Solubility (1 mmole in 2 ml DMF):Clearly Soluble
- Assay (acidimetric):≥97,0%
- Water (K. F.):≤1,00%
- Storage:Store below 30 °C
Frequently Bought Together