Order Entry
Italy
ContactUsLinkComponent
Sudan IV, red brown, powder C.I. 26105
Sudan IV, red brown, powder C.I. 26105
Catalog # ICNA0215264825
Supplier:  MP Biomedicals
CAS Number:  
Sudan IV, red brown, powder C.I. 26105
Catalog # ICNA0215264825
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Oil Red, 1-((2-Methyl-4-(o-tolyldiazenyl)phenyl)diazenyl)naphthalen-2-ol, Scarlet Red, Oil Red BB, 1-({2-Methyl-4-[(2-methylphenyl)diazenyl]-phenyl}diazenyl)-2-naphthol, Scarlet R, Biebrich scarlet R, Lipid Crimson, Scarlet Red Scharlach, Oil Red IV, Fat Red B, Solvent Red 24, 1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphthol
 :
 :380.45 g/mol
CAS Number:85-83-6
Formula:C₂₄H₂₀N₄O
MDL Number:MFCD00003893
Melting Pt:188
Merck Index:12,08538