CytoScint™ ES is a complete, ready-to-use environmentally safe liquid scintillation solution designed for 'one-step' counting of filters, thin-layer chromatography scrapings, and samples solubilised in Hyamine® Hydroxide.
- Environmentally safe
- One step filter counting
- Non-toxic and non-flammable
- Stable counting performance
- No waiting on chemiluminescence
- High quench resistance
For most samples containing minimal amounts of water (0 to 15%), it will result in increased count recovery and maximum sample solubilisation.