
Acid red 52, powder C.I. 45100
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:3520-42-1
:
Formula:C₂₇H₃₀N₂O₇S₂
MDL Number:MFCD00010180
:580.65 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test :Passes
- UV/VIS: λmax m (0.1N NaOH):565 ± 2 nm
Specification Test Results
Frequently Bought Together