Quercetin dihydrate, greenish-yellow powder
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate
CAS Number:6151-25-3
:
Formula:C₁₅H₁₄O₉
MDL Number:MFCD00149487
Molecular Weight:338.268 g/mol
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- UV/VIS: λmax (ethanol):255 - 261, 372 - 37 nm
- Moisture:9 - 12%
Specification Test Results
Frequently Bought Together