Phaseolus vulgaris (Red Kidney Bean) Phytohemagglutin-leukoagglutinin, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Reconstitution: Reconstitute with water to a final concentration of 1 mg/ml (use 1 ml for the 1 mg size or 5 ml for the 5 mg size) for a final solution of 1 mg/ml in 0,40 mM sodium phosphate, 6 mM NaCl, pH 7,45. Reconstitute with water to a final concentration of 25 mg/ml (0,2 ml water for the 5 mg size). This concentration of the lectin has been used for iontophoresis and neuron transport.
Carbohydrate Specificity: PHA-L recognizes a complex branched chain oligosaccharide structure.
Specifications
- Storage conditions:Store at -0 °C, Store solutions sterile, Filtered and frozen in aliquots. Avoid freeze-thaw cycles, Clarify by centrifugation
- Form:White Solid
- Protein synonyms:PHA-P|PHA|Phaseolus vulgaris agglutinin|PHA-L|Lectin from Phaseolus vulgaris
- Protein/peptide name:Phytohemagglutin-leukoagglutinin
Specifications
Frequently Bought Together