Order Entry
Italy
ContactUsLinkComponent
 
5-Fluoro-2'-deoxyuridine (floxuridine) ≥98%, white powder
 
5-Fluoro-2'-deoxyuridine (floxuridine) ≥98%, white powder

 

  • Identity Test:
    Passes
  • Purity:
    ≥98%
  • FTIR:
    Conforms to Standard
  • Melting Point:
    142 - 155 °C
  • UV/VIS: l max (water):
    266 - 270 nm

 

Frequently Bought Together