Paraformaldehyde, powder
Supplier: MP Biomedicals
Jump to:
About this item
Soluble in water (hot water - ~37% w/v; slowly soluble in cold water). Aqueous solutions will develop formaldehyde over time. Insoluble in alcohol or ether.
Synonyms: Polyoxymethylene
- CAS Number:30525-89-4
- Formula:(CH₂O)n
- MDL Number:MFCD00133991
- UN:2213
- ADR:4.1,III
3 Options Available Below
Product Details & Documents
Soluble in water (hot water - ~37% w/v; slowly soluble in cold water). Aqueous solutions will develop formaldehyde over time. Insoluble in alcohol or ether.
Synonyms: Polyoxymethylene
- CAS Number:30525-89-4
- Formula:(CH₂O)n
- MDL Number:MFCD00133991
- UN:2213
- ADR:4.1,III
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



