
Tetracycline hydrochloride, yellow powder
:64-75-5
:
:C₂₂H₂₅ClN₂O₈
:MFCD00078142
:480.9 g/mol
:GHS07
:Freezer
- Identity Test:Passes
- Potency:≥900 µg/mg
- FTIR:Conforms to Standard
- Melting Point:215 - 225 °C
- UV/Vis: λmax (1N HCl):219, 271, 357 ± 5 nm
- pH (1% aq soln):1.8 - 2.8
- Moisture Content:≤2%
- Optical Rotation:-239 ° to -258 ° (c=0.01, 0.1 N HCl)
Frequently Bought Together