Order Entry
Italy
ContactUsLinkComponent
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-glucopyranose ≥95%
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-glucopyranose ≥95%
Supplier:  Cayman Chemical
Certificates
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-glucopyranose ≥95%
Supplier:  Cayman Chemical

Specification Test Results

  • Purity:
    ≥95%
  • Formal Name:
    2-(acetylamino)-2-deoxy-β-D-glucopyranose, 1,3,4,6-tetraacetate
  • Synonyms:
    N-Acetyl-β-D-Glucosamine tetraacetate, NSC 224432
  • Formulation:
    A Crystalline Solid
  • SMILES:
    CC(OC[C@H]([C@H]([C@@H]([C@H]1NC(C)=O)OC(C)=O)OC(C)=O)O[C@H]1OC(C)=O)=O
  • InChI Code:
    1S/C16H23NO10/c1-7(18)17-13-15(25-10(4)21)14(24-9(3)20)12(6-23-8(2)19)27-16(13)26-11(5)22/h12-16H,6H2,1-5H3,(H,17,18)/t12-,13-,14-,15-,16-/m1/s1
  • InChI Key:
    OVPIZHVSWNOZMN-OXGONZEZSA-N
  • Storage:
    −20 °C
  • Shipping:
    Room temperature in continental US; may vary elsewhere

Specification Test Results

Frequently Bought Together