D-erythro-Sphingosine C-17 ≥98%
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:6918-48-5
:
Formula:C₁₇H₃₅NO₂
MDL Number:MFCD18427933
:285.472 g/mol
Specification Test Results
- Purity:≥98%
- Formal Name:2S-amino-4E-heptadecene-1,3R-diol
- Formulation:A Crystalline Solid
- SMILES:OC[C@H](N)[C@H](O)/C=C/CCCCCCCCCCCC
- InChI Code:1S/C17H35NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-17(20)16(18)15-19/h13-14,16-17,19-20H,2-12,15,18H2,1H3/b14-13+/t16-,17+/m0/s1
- InChI Key:RBEJCQPPFCKTRZ-LHMZYYNSSA-N
- Storage:−20 °C
- Shipping:Room temperature in continental US; may vary elsewhere
- Stability:2 years from the QC date provided on the Certificate of Analysis, when stored properly
Specification Test Results
Frequently Bought Together