


L-Selenomethionine ≥97% (by HPLC), solid, Calbiochem®, Millipore®
Nome del fornitore: Merck Millipore (Calbiochem)
Certificati
Informazioni su questo articolo
Numero CAS:3211-76-5
:
Formula:C₅H₁₁NO₂Se
Numero MDL:MFCD00037210
:196.11 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ100
UN:3283
:GHS06
:GHS08
:GHS09
temperatura di conservazione:Ambient
Risultati dei test sulle specifiche
- Synonyms:SeMet
- Form:White to Off-white Solid
- Purity:≥98% by HPLC
- Contaminants (Heavy Metals):≤10 ppm
- Contaminants (Selenium):≥39,5%
- Ship Code:Ambient Temperature only
- Toxicity:Carcinogenic / Teratogenic
- Storage:15 to 30 °C
- Do not freeze:Ok to freeze
- Special Instructions:Unstable in solution; reconstitute just prior to use
- Solubility:H2O (50 mg/ml)
Risultati dei test sulle specifiche
Frequently Bought Together