2'-Deoxycytidine hydrochloride, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:3992-42-5
:
Formula:C₉H₁₄ClN₃O₄
MDL Number:MFCD00012840
:263.68 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
Specification Test Results
Frequently Bought Together