Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥97%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:19817-92-6
:
Formula:C₉H₁₂N₂Na₃O₁₅P₃
MDL Number:MFCD00044310
:550.09 g/mol
Specification Test Results
- Identity Test:Passes
- Purity:≥97%
- FTIR:Conforms to Standard
- OD 250/260:0.74 - 0.79
- OD 280/260:0.32 - 0.40
Specification Test Results
Frequently Bought Together