Order Entry
Italy
ContactUsLinkComponent
Human AXL ELISA kit
Human AXL ELISA kit
Catalog # BIRBORB138059-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human AXL ELISA kit
Catalog # BIRBORB138059-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human AXL ELISA kit, 96-well
  • Detection range:
    62,5 - 4000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human AXL ELISA Kit is used for quantitative detection of human AXL in cell culture supernates, serum and plasma (heparin, EDTA).

Tyrosine-protein kinase receptor UFO is an enzyme that in humans is encoded by the AXL gene. The protein encoded by this gene is a member of the receptor tyrosine kinase subfamily.

Axl is a chronic myelogenous leukemia-associated oncogene and also associated with colon cancer and melanoma. It is in close vicinity to the BCL3 oncogene, which is at 19q13.1-q13.2. The AXL gene is evolutionarily conserved between vertebrate species. This gene has two different alternatively spliced transcript variants.