DL-Arabinose, white powder
Catalog # ICNA0210070010
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:150.13 g/mol
Storage Temperature:Ambient
CAS Number:147-81-9
:20235-19-2
Formula:C₅H₁₀O₅
MDL Number:MFCD00135867