Order Entry
Italy
ContactUsLinkComponent
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) ≥97%, colorless solid
Corrosive
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) ≥97%, colorless solid
Supplier:  MP Biomedicals
Certificates
Corrosive
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) ≥97%, colorless solid
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥97%
  • FTIR:
    Conforms to Standard
  • pH (0.5% aq soln):
    11 - 14

Specification Test Results

Frequently Bought Together