Anti-FHL3 Rabbit Polyclonal Antibody
Catalog # BSBTPB10064
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Four and a half LIM domains 3
- Antigen symbol:FHL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2275
- Antigen synonyms:SLIM2
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Four and a half LIM domains protein 3(FHL3) detection. Tested with WB in Human;Rat.
Type: Primary
Antigen: FHL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat