Anti-ITGA241 Mouse Monoclonal Antibody (PE) [clone: TEA1/41]
Catalog # ABNOMAB13689
Nome del fornitore: Abnova
Specifiche
- Antibody type:Primary
- Antigen symbol:ITGA241
- Clonality:Monoclonal
- Clone:TEA1/41
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:3673
- Size:100 reactions
- Pk:100 Assays
Specifiche
Informazioni su questo articolo
Mouse monoclonal antibody raised against human ITGA2.
Type: Primary
Antigen: ITGA241
Clonality: Monoclonal
Clone: TEA1/41
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human