Anti-CMV gB Mouse Monoclonal Antibody [clone: P1201]
ABNOMAB13514
: Abnova
- Antibody type:Primary
- Antigen name:Cytomegalovirus gB
- Antigen symbol:CMV gB
- Clonality:Monoclonal
- Clone:P1201
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Cytomegalovirus
- Size:500 μg
- Pk:500 µG
Mouse monoclonal antibody raised against cytomegalovirus (CMV) gB.
Type: Primary
Antigen: CMV gB
Clonality: Monoclonal
Clone: P1201
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Cytomegalovirus (CMV)