Order Entry
Italy
ContactUsLinkComponent
 
Armenian Hamster IgG Isotype Control [clone: PIP] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Catalog # 65-4888-U100
Supplier:  Tonbo Biosciences
Armenian Hamster IgG Isotype Control [clone: PIP] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Catalog # 65-4888-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    PIP
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Flow cytometry:
    Yes
  • Host:
    Armenian hamster
  • Isotype:
    IgG
  • Storage temperature:
    +2 °C to +8 °C, protected from light
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

The PIP immunoglobulin is useful as an isotype-matched control. The PIP immunolglobulin is specific for Glutathione S-transferase and can be used as an isotype control for Armenian Hamster IgG antibodies.

Type: Control
Antigen: Isotype Control
Clonality: Monoclonal
Clone: PIP
Conjugation: PerCP-Cy5.5
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: N/A