Capillary columns, Integra-Guard®
Supplier: Restek
Certificates
About this item
Integra-Guard® columns are designed for leak-free connection between the guard column and the analytical column. The guard column section is marked separately from the analytical column, using high-temperature string.
- No leaks for a more robust method
- No column connections
- Easy for maintenance
- No peak distortions
Frequently Bought Together