Polystyrene standards for Gel Permeation Chromatography (GPC)
Molecular weight standard for gel permeation chromatography (GPC).
- Powders
- Odourless
CAS No.: 9003-53-6
MDL No.: MFCD00084450
- CAS No.:9003-53-6
- MDL No.:MFCD00084450
- Formula:C₈H₈
- Description:Polystyrene MW standard
- Physical form:Powder
- Odour/scent:Odourless
Frequently Bought Together