Stirring Rods, Thermo Fisher Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
The fused quartz rods are manufactured by fusing naturally occurring crystalline silica.
- Fused quartz has high chemical purity, high corrosion resistance and high melting point
- Quartz is extremely hard, with a low coefficient of thermal expansion, excellent electrical insulation qualities and optical transmission from ultra-violet to infra-red
Frequently Bought Together