Order Entry
Italy
ContactUsLinkComponent
Dibismuth trisulphate 99%, pure
Dibismuth trisulphate 99%, pure
Certificates
undefined
Dibismuth trisulphate 99%, pure

Specification Test Results

  • Appearance:
    White Powder
  • Assay:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together