Order Entry
Italy
ContactUsLinkComponent
5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine 97%
Irritant
5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine 97%
Catalog # ACRO453590050
CAS Number:  
undefined
Irritant
5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine 97%
Catalog # ACRO453590050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
Melting Pt:94…97 °C
 :319.23 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:365996-06-1
Formula:C₁₁H₁₅BrN₂O₂S
MDL Number:MFCD09951958