Stachyose hydrate, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54261-98-2
:
Formula:C₂₄H₄₂O₂₁·xH₂O
MDL Number:MFCD00149457
:666.58 g/mol (anhy)
Melting Pt:95…105 °C (dec.)
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow to light beige Powder
- Infrared spectrum:Conforms
- Water:≤12 %
Specification Test Results
Frequently Bought Together