Order Entry
Italy
ContactUsLinkComponent
 
N(α)-Acetyl-L-tryptophan 97%
 
undefined
N(α)-Acetyl-L-tryptophan 97%

 

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥96.0 %
  • Loss on drying:
    ≤0.3 %
  • Specific optical rotation:
    +23° to +28° (20°C, 589 nm) (c=1, EtOH)

 

 

Frequently Bought Together