
Sulphuric acid 25% in water for analysis
Catalog # ACRO451050010
Supplier: Thermo Fisher Scientific
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Appearance:Clear liquid
- Color scale:≤10 APHA
- Residue after ignition:≤0.0005 %
- Titration with NaOH:≥25 %
- Chloride (Cl):≤0.5 ppm
- Nitrate (NO3):≤0.2 ppm
- Aluminium (Al):≤0.05 ppm
- Ammonium (NH4):≤2 ppm
- Arsenic (As):≤0.01 ppm
- Beryllium (Be):≤0.02 ppm
- Cadmium (Cd):≤0.02 ppm
- Calcium (Ca):≤0.2 ppm
- Chromium (Cr):≤0.05 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.01 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.02 ppm
- Lithium (Li):≤0.02 ppm
- Magnesium (Mg):≤0.05 ppm
- Manganese (Mn):≤0.01 ppm
- Nickel (Ni):≤0.02 ppm
- Potassium (K):≤0.1 ppm
- Silver (Ag):≤0.02 ppm
- Sodium (Na):≤0.5 ppm
- Strontium (Sr):≤0.02 ppm
- Zinc (Zn):≤0.1 ppm
- Heavy metals (as Pb):≤1 ppm
- KMnO4-reducing substance:≤0.0002 % (as SO2)
Specification Test Results
About this item
:
:98.08 g/mol
Boiling Pt:∼100 °C (1013 hPa)
Density:1,1 to 1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2796
ADR:8,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589