
6,7-Dihydro-5H-pyrrolo[3,4-b]pyridine dihydrochloride 97%
Catalog # ACRO449900010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to green Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 %
Specification Test Results
About this item
:
:193.0700 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:147740-02-1
Formula:C₇H₁₀Cl₂N₂
MDL Number:MFCD09607967