Order Entry
Italy
ContactUsLinkComponent
4'-Chloro-2'-hydroxyacetophenone 97%
Irritant
4'-Chloro-2'-hydroxyacetophenone 97%
Certificates
undefined
Irritant
4'-Chloro-2'-hydroxyacetophenone 97%

Specification Test Results

  • Appearance:
    White to light yellow Crystalline mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    27 to 30 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together