Order Entry
Italy
ContactUsLinkComponent
2,6-Dibromonaphthalene 99%
2,6-Dibromonaphthalene 99%
Certificates
undefined
2,6-Dibromonaphthalene 99%

Specification Test Results

  • Appearance:
    White to beige Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    162 to 167 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together