3-Amino-5-chloropyridine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22353-34-0
:
Formula:C₅H₅ClN₂
MDL Number:MFCD03701386
:128.56 g/mol
Melting Pt:51…56 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS05
:GHS06
Specification Test Results
- Appearance:Cream to beige to light brown Powder
- Infrared spectrum:Conforms
- Melting point:71 to 75 °C
- GC:≥94 %
Specification Test Results
Frequently Bought Together