

4,4-Dimethylcyclohexanone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4255-62-3
:
Formula:C₈H₁₄O
MDL Number:MFCD00234965
:126.2 g/mol
Boiling Pt:73 °C (14 mmHg)
Melting Pt:41…43 °C
Flash Pt:62 °C
Storage Temperature:Ambient
UN:1325
ADR:4.1,II
:GHS02
:GHS07
Specification Test Results
- Appearance:White to tan Crystals or solid
- Infrared spectrum:Conforms
- Melting point:41 to 45 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together