Order Entry
Italy
ContactUsLinkComponent
Ammonium iodide 99%, extra pure
Irritant
Ammonium iodide 99%, extra pure
Certificates
undefined
Irritant
Ammonium iodide 99%, extra pure
Certificates

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder (May turn yellow on storage)
  • Titration Argentometric:
    98.5 to 101.5 %
  • Residue after ignition:
    ≤0.05 %
  • Heavy metals (as Pb):
    ≤0.001 %

Specification Test Results

Related Information

Frequently Bought Together