Order Entry
Italy
ContactUsLinkComponent
Sodium fluoride for biochemistry
Toxic
Sodium fluoride for biochemistry
Catalog # ACRO447355000
CAS Number:  
undefined
Toxic
Sodium fluoride for biochemistry
Catalog # ACRO447355000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Titration:
    ≥98.5 %
  • Chloride (Cl):
    ≤0.005 %
  • Sulfate (SO4):
    ≤0.03 %
  • Iron (Fe):
    ≤0.003 %
  • Potassium (K):
    ≤0.02 %
  • Heavy metals (as Pb):
    ≤0.003 %
  • Insoluble matter:
    ≤0.1 %

Specification Test Results

About this item

 :
 :41.99 g/mol
Boiling Pt:1700 °C (1013 hPa)
Melting Pt:993 °C
Density:2,8 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1690
ADR:6.1,III
 :GHS06
CAS Number:7681-49-4
Formula:NaF
MDL Number:MFCD00003524