Order Entry
Italy
ContactUsLinkComponent
4-(Trifluoromethyl)-1H-pyrazole 97%
Irritant
4-(Trifluoromethyl)-1H-pyrazole 97%
Catalog # ACRO446490010
CAS Number:  
undefined
Irritant
4-(Trifluoromethyl)-1H-pyrazole 97%
Catalog # ACRO446490010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to yellow Crystalline solid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤2.5 %
  • GC:
    ≥96 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

About this item

 :
 :136.077 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:52222-73-8
Formula:C₄H₃F₃N₂
MDL Number:MFCD11226572