
Aluminium nitrate nonahydrate ACS
Catalog # ACRO446340010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Appearance:Colorless to white Crystalline powder or crystals
- Titration Complexometric:98.0 to 102.0 %
- Chloride (Cl):≤0.001 %
- Sulfate (SO4):≤0.005 %
- Calcium (Ca):≤0.005 %
- Iron (Fe):≤0.002 %
- Magnesium (Mg):≤0.001 %
- Potassium (K):≤0.002 %
- Sodium (Na):≤0.005 %
- Heavy metals (ICP-OES):≤0.001 %
- Insoluble matter:≤0.005 %
Specification Test Results
About this item
:
:375.13 g/mol
Melting Pt:73 °C
Density:1,72 g/cm³ (20 °C)
ADR:5.1,III
:GHS05
Storage Temperature:Ambient
CAS Number:7784-27-2
Formula:Al(NO₃)₃·9H₂O
MDL Number:MFCD00149132