Order Entry
Italy
ContactUsLinkComponent
3,3-Dimethoxypropiononitrile 90%
Irritant
3,3-Dimethoxypropiononitrile 90%
Certificates
undefined
Irritant
3,3-Dimethoxypropiononitrile 90%

Specification Test Results

  • Appearance:
    Colorless to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4120 to 1.4160 (20°C, 589 nm)
  • GC:
    ≥88 %

Specification Test Results

Related Information

Frequently Bought Together